Learn More
Strem, An Ascensus Company CAS 300344-02-9. 500mg. 26-Diisopropylphenylimidoneophylidene[racemic-BIPHEN]molybdenum(VI) min. 97% rac-SCHROCK-HOVEYDA CATALYST. MFCD01862462
SDP
Supplier:  Strem, An Ascensus Company 421212500MG
Encompass
CAS 300344-02-9. 500mg. 26-Diisopropylphenylimidoneophylidene[racemic-BIPHEN]molybdenum(VI) min. 97% rac-SCHROCK-HOVEYDA CATALYST. MFCD01862462. Molecular Weight 755.93. Molecular Formula Mo(C10H12)(C12H17N)(C24H32O2). Color/form red xtl. Strem 42-1212. http//www.strem.com/catalog/v/42-1212/
Catalog No. 50-211-0813