Learn More
Strem, An Ascensus Company CAS# 139220-25-0. 2g. 2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide) SCHROCK'S CATALYST. MFCD00269857
SDP
Catalog No. 502045310
Encompass
CAS# 139220-25-0. 2g. 2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide) SCHROCK'S CATALYST. MFCD00269857. Molecular Weight: 765.53. Molecular Formula: Mo(C10H12)(C12H17N)[OC(CH3)(CF3)2]2. Color/form: yellow to orange pwdr. Strem# 42-1205. http://www.strem.com/catalog/v/42-1205/
Catalog No. 50-204-5310 Supplier Strem, An Ascensus Company Supplier No. 4212052G
CAS# 139220-25-0. 2g. 2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide) SCHROCK'S CATALYST. MFCD00269857. Molecular Weight: 765.53. Molecular Formula: Mo(C10H12)(C12H17N)[OC(CH3)(CF3)2]2. Color/form: yellow to orange pwdr. Strem# 42-1205. http://www.strem.com/catalog/v/42-1205/