Learn More
Med Vet International Miltex Vantage Schroeder Tenaculum Forceps, 9.5in., Straight
Catalog No. 502829250
Encompass_Preferred
The Miltex Vantage V930-965 Schroeder Tenaculum Forceps are made of mid-grade stainless steel and have a length of 9.5in. They have 1x1 prongs with a ratcheted, finger ring. This surgical forcep is commonly used to grasp, hold, and move vessels and arteries during surgical procedures. It may also be used in OB/GYN procedures involving manipulation of the cervix or uterus, and vaginal or abdominal hysterectomy.
Catalog No. 50-282-9250 Supplier Med Vet International Supplier No. MILV930965
The Miltex Vantage V930-965 Schroeder Tenaculum Forceps are made of mid-grade stainless steel and have a length of 9.5in. They have 1x1 prongs with a ratcheted, finger ring. This surgical forcep is commonly used to grasp, hold, and move vessels and arteries during surgical procedures. It may also be used in OB/GYN procedures involving manipulation of the cervix or uterus, and vaginal or abdominal hysterectomy.