Learn More
Cayman Chemical Gomisin J, Purity: ≥98%, CAS: 66280-25-9, 10 mg
SDP
Catalog No. 502988584
Encompass_Preferred
A lignan; inhibits HIV-1 IIIB replication in H9 T cells (EC50 = 1.5 µg/ml); decreases LPS-induced NO production and p38, ERK, and JNK phosphorylation in RAW 264.7 cells; induces relaxation of isolated, precontracted endothelium-intact rat aortic rings at 3, 10, and 30 µg/ml; cytotoxic to 13 cancer cell lines at 30 µg/ml;
Catalog No. 50-298-8584 Supplier Cayman Chemical Supplier No. 3021310 MG
A lignan; inhibits HIV-1 IIIB replication in H9 T cells (EC50 = 1.5 µg/ml); decreases LPS-induced NO production and p38, ERK, and JNK phosphorylation in RAW 264.7 cells; induces relaxation of isolated, precontracted endothelium-intact rat aortic rings at 3, 10, and 30 µg/ml; cytotoxic to 13 cancer cell lines at 30 µg/ml;