Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciencell Research Laboratories Schwann Cell Medium, 500 ml, complete medium, sterile liquid, 5% FBS, buffered with HEPES and bicarbonate, pH 7.4, for normal Schwann cell growth

Catalog No. NC0017717
Encompass

ScienCell Research Laboratories' Schwann Cell Medium (SCM) is designed for the optimal in vitro growth and support of normal Schwann cells. This complete, sterile liquid medium is carefully formulated to maintain a stable pH and provide essential nutrients for cell development. It includes various components such as amino acids, vitamins, and growth factors to enhance cell stability and efficacy during culture.

  • Complete medium needs supplementation for optimal use.
  • Volume: 500 ml.
  • FBS Concentration is 5%.
  • Buffered with HEPES and bicarbonate.
  • Maintains pH of 7.4 in 5% CO2/95% air incubator.
  • Sterile liquid medium.
  • Includes essential amino acids, vitamins, trace minerals.
  • Targeted for normal Schwann cell growth.
  • Storage for basal medium at 4°C; supplements at -20°C.
  • Ships at room temperature for basal medium; dry ice for supplements.
  • Intended for research use only.

Catalog No. NC0017717 Supplier Sciencell Research Laboratories Supplier No. 1701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ScienCell Research Laboratories' Schwann Cell Medium (SCM) is designed for the optimal in vitro growth and support of normal Schwann cells. This complete, sterile liquid medium is carefully formulated to maintain a stable pH and provide essential nutrients for cell development. It includes various components such as amino acids, vitamins, and growth factors to enhance cell stability and efficacy during culture.

  • Complete medium needs supplementation for optimal use.
  • Volume: 500 ml.
  • FBS Concentration is 5%.
  • Buffered with HEPES and bicarbonate.
  • Maintains pH of 7.4 in 5% CO2/95% air incubator.
  • Sterile liquid medium.
  • Includes essential amino acids, vitamins, trace minerals.
  • Targeted for normal Schwann cell growth.
  • Storage for basal medium at 4°C; supplements at -20°C.
  • Ships at room temperature for basal medium; dry ice for supplements.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.