Learn More
Fine Science Tools Schwartz Micro Serrefines, Jaw: 10 x 1.75mm, Sharp Bend, 26 mm
SDP
Catalog No. NC9248218
Encompass
Product: Schwartz Micro Serrefine - Sharp Bend/26mm/ Shape: Sharp Bend/ Jaw Dimensions: 10 x 1.75mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 26mm/ Serrated: No/ Unit: Each
Catalog No. NC9248218 Supplier Fine Science Tools Supplier No. 1805203
Product: Schwartz Micro Serrefine - Sharp Bend/26mm/ Shape: Sharp Bend/ Jaw Dimensions: 10 x 1.75mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 26mm/ Serrated: No/ Unit: Each