Learn More
Roboz Surgical SCHWARTZ CLIP STRONG ANGLE
SDP
Catalog No. NC9701416
Encompass
Schwartz Temporary Clip; Strong Angle, Smooth; 795 Grams Pressure; 1.7mm Jaw Width; 8mm Jaw Length; 1 in. Clip Length Not for human use
Catalog No. NC9701416 Supplier Roboz Surgical Supplier No. RS5459
Schwartz Temporary Clip; Strong Angle, Smooth; 795 Grams Pressure; 1.7mm Jaw Width; 8mm Jaw Length; 1 in. Clip Length Not for human use