Description
- Using the applying forceps, 52106-73, surgical clips can easily be position on the vessels. The locking forceps allows for easy, precise, and secure grasping of the micro surgical clips. Simply grasp the clip with the forceps to open the jaw, slid it over the vessels and release.
- To remove surgical clips, simply perform the procedure in reverse.
- This procedure does not compromise the vessels integrity.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | 1 in. |
| Model | Straight, Smooth |