Learn More
Roboz Surgical SCHWARTZ TEMP CLIP
SDP
Catalog No. NC9956119
Encompass
Schwartz Temporary Clip; Straight, Smooth; 795 Grams Pressure; 1.7mm Jaw Width; 8mm Jaw Length; 1 in. Clip Length Not for human use
Catalog No. NC9956119 Supplier Roboz Surgical Supplier No. RS5452
Schwartz Temporary Clip; Straight, Smooth; 795 Grams Pressure; 1.7mm Jaw Width; 8mm Jaw Length; 1 in. Clip Length Not for human use