Learn More
Bellco Glass, Inc. Sci/ERA 10 Deck, 230V Fixed Frame - 50 Position
SDP
Catalog No. 501945939
Encompass_Preferred
The Bellco Sci/ERA Cell Production Roller Apparatus has been designed for production of mammalian cells, viruses and cell products. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. All units feature a solid state control with heavy-duty DC motor and digital tachometer. All Position Drives are for use with plastic roller bottles to prevent slipping of the light-weight bottles. A Stand-By Drive and Alarm System provides motion and power failure alarms and has a backup drive system that automatically drives the unit should the original fail. MADE IN THE USA
Catalog No. 50-194-5939 Supplier Bellco Glass, Inc. Supplier No. 763082510
Add to Cart
Add to Cart
Request a Quote
The Bellco Sci/ERA Cell Production Roller Apparatus has been designed for production of mammalian cells, viruses and cell products. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. All units feature a solid state control with heavy-duty DC motor and digital tachometer. All Position Drives are for use with plastic roller bottles to prevent slipping of the light-weight bottles. A Stand-By Drive and Alarm System provides motion and power failure alarms and has a backup drive system that automatically drives the unit should the original fail. MADE IN THE USA