Learn More
Andwin Scientific SCI INDUSTRIES VORTEX GENIE II
SDP
Catalog No. NC9612654
Encompass_Preferred
The Vortex-Genie 2 mixes or shakes liquids in all types of container with efficient controllable speed and precision. This product features a sturdy metal housing that gives a stable platform for all kinds of mixing. It also has a timer that will turn the unit off after a certain time. This timer can be activated with touch and hands-free. This product provides reproducible results with an accuracy of 5% and repeatability of 1%. Other accessories or attachments that can expand the uses of this product are sold separately.
Catalog No. NC9612654 Supplier Andwin Scientific Supplier No. G560
The Vortex-Genie 2 mixes or shakes liquids in all types of container with efficient controllable speed and precision. This product features a sturdy metal housing that gives a stable platform for all kinds of mixing. It also has a timer that will turn the unit off after a certain time. This timer can be activated with touch and hands-free. This product provides reproducible results with an accuracy of 5% and repeatability of 1%. Other accessories or attachments that can expand the uses of this product are sold separately.