Learn More
Cayman Chemical SCIADOPITYSIN 10MG
SDP
Catalog No. 502939010
Encompass_Preferred
A biflavonoid with diverse biological activities; reduces Aβ42-induced cytotoxicity in PC12 cells (EC50 = 9.84 μM); decreases methylglyoxal-induced insulin secretion, production of ROS, cardiolipin peroxidation, and cytotoxicity in RIN-m5F pancreatic β-cells from 0.1-10 μM; inhibits P-gp (IC50 = 53.42 μM) and increases cellular toxicity of paraquat and paclitaxel in MDR1-MDCKII cells; inhibits RANKL-induced mRNA expression of the osteoclast-specific genes CTSK, TRAP, and MMP-9, activation of NF-κB, and osteoclastogenesis in a mouse model of LPS-induced bone loss
Catalog No. 50-293-9010 Supplier Cayman Chemical Supplier No. 2790010 MG
A biflavonoid with diverse biological activities; reduces Aβ42-induced cytotoxicity in PC12 cells (EC50 = 9.84 μM); decreases methylglyoxal-induced insulin secretion, production of ROS, cardiolipin peroxidation, and cytotoxicity in RIN-m5F pancreatic β-cells from 0.1-10 μM; inhibits P-gp (IC50 = 53.42 μM) and increases cellular toxicity of paraquat and paclitaxel in MDR1-MDCKII cells; inhibits RANKL-induced mRNA expression of the osteoclast-specific genes CTSK, TRAP, and MMP-9, activation of NF-κB, and osteoclastogenesis in a mouse model of LPS-induced bone loss