Learn More
TARGETMOL CHEMICALS INC SCIADOPITYSIN 10MG
SDP
Catalog No. 502579709
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sciadopitysin may prevent the development of diabetic osteopathy it exerts its therapeutic effects via upregulation of mitochondrial biogenesis.Sciadopitysin shows protective effects on antimycin A-induced toxicity in osteoblastic MC3T3-E1 cells it may reduce or prevent osteoblasts degeneration.Sciadopitysin can inhibit the A(beta) aggregation and reduce A(beta)-induced toxicity in the primary cortical neurons. purity: 99%
Catalog No. 50-257-9709 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S212910MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sciadopitysin may prevent the development of diabetic osteopathy it exerts its therapeutic effects via upregulation of mitochondrial biogenesis.Sciadopitysin shows protective effects on antimycin A-induced toxicity in osteoblastic MC3T3-E1 cells it may reduce or prevent osteoblasts degeneration.Sciadopitysin can inhibit the A(beta) aggregation and reduce A(beta)-induced toxicity in the primary cortical neurons. purity: 99%