Learn More
Medchemexpress LLC HY-N2119 5mg Medchemexpress, Sciadopitysin CAS:521-34-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21195MG
Encompass_Preferred
Medchemexpress, HY-N2119 5mg Sciadopitysin CAS:521-34-6 Sciadopitysin is a type of biflavonoids in leaves from ginkgo biloba. Sciadopitysi inhibits RANKL-induced osteoclastogenesis and bone loss by inhibiting NF-κB activation and reducing the expression of c-Fos and NFATc1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0368