Learn More
PARKER HANNIFIN CORP SCICON CND SENSR 3/8 BARB 5PK
SDP
Catalog No. NC0074480
Encompass
3/8" Barb for tube size range ID 0.31" to 0.38".,8 LPM, 60 PSI. SciLog� SciCon� pre-calibrated single-use conductivity sensors are available in 5 sizes, pre-programmed and barcoded with a unique ID for full traceability and easy data collection. Calibration and identity data are stored on the sensor's chip.
Catalog No. NC0074480 Supplier PARKER HANNIFIN CORP Supplier No. 080594PSX5
3/8" Barb for tube size range ID 0.31" to 0.38".,8 LPM, 60 PSI. SciLog� SciCon� pre-calibrated single-use conductivity sensors are available in 5 sizes, pre-programmed and barcoded with a unique ID for full traceability and easy data collection. Calibration and identity data are stored on the sensor's chip.