Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Component Kit
GREENER_CHOICESCI_ED

Catalog No. S89516 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89516 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89516 Supplier Supertek™ Supplier No. PH96022S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Looking to do more experiments? Find what you need in this Component Kit! These basic science components are a great way to expand our Telegraph Kit, Simple Circuit, Conduction Experiment, and Color Theory. Perfect for group experiments and homeschool!

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 1 single throw knife switch
    • 1 double throw knife switch
    • 2 light bulb holders
    • 2 AA Cell Battery Holder
    • 10 Light Bulbs
    • 10 Alligator Connector Cords
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Science Experiments
For Use With (Equipment) Telegraph Kit, Simple Circuit, Conduction Experiment, and Color Theory
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.