Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Conduction Experiments
GREENER_CHOICESCI_ED

Catalog No. S89514 Shop All Supertek Scientific Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S89514 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89514 Supplier Supertek™ Supplier No. PH96020S3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Does wood conduct electricity? How about a penny? Make your own Conduction Experiment and explore what substances are conductors and which are insulators. Hint, most substances can be classified as one or the other.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 2 D cell battery holders
    • 1 light bulb
    • 1 light bulb holder
    • 4 alligator connector cords
    • 1 toothpick
    • 1 Penny
    • 2 Paper Clips
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Make Conduction Experiment
Includes Reusable Plastic Snap Box, D Cell Battery Holders, Light Bulb, Light Bulb Holder, Alligator Connector Cords, Toothpick, Penny, Paper Clips, Exploration Guide
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.