Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Culture Cube/Petri Dishes with Agar
GREENER_CHOICESCI_ED

Catalog No. S89525 Shop All Supertek Scientific Products
Change view
Click to view available options
Diameter (Metric):
35 mm and 90 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S89525 35 mm and 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89525 Supplier Supertek™ Supplier No. PH96004S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover the fun of microbiology with our Culture Cube/Petri Dishes with Agar! Science will come alive literally as you explore common household bacteria. Perfect for group experiments and homeschool!

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 8 large petri dishes
    • 8 small petri dishes
    • 8 swabs
    • 1 packet nutrient agar
    • 1 Exploration Guide
  • Ages 8+

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Single-use
Material Plastic
Shape Round
Sterility Non-sterile
Surface Treatment Non-treated
Type Petri Dish
For Use With (Application) Exploring Common Household Bacteria
Includes Reusable Plastic Snap Box, 8 Large Petri Dishes, 8 Small Petri Dishes, 8 Swabs, 1 Packet Nutrient Agar, 1 Exploration Guide
Height (Metric) 15 mm
Diameter (Metric) 35 mm and 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.