Learn More
Description
- Perfect for Group Experiments
- Perfect for Homeschooling
- Save space when storing in reusable plastic snap box
- Includes:
- 1 Reusable plastic snap box
- 1 Flexible Mirror
- 1 Cylindrical Double Concave Lens
- 1 Cylindrical Double Convex Lens
- 1 Small Equilateral Prism
- 1 Small Mirror
- 1 Laser
- 4 Cards
- 1 Exploration Guide
- Ages 7+
Specifications
Specifications
| Grade | 1 |
| Age | 7 |
| For Use With (Application) | Explore Reflection, Refraction, and the Physics of Light |
| Includes | Reusable Plastic Snap Box, Flexible Mirror, Cylindrical Double Concave Lens, Cylindrical Double Convex Lens, Small Equilateral Prism, Small Mirror, Laser, 4 Cards, Exploration Guide |
| Class Size | 1 Lab Group |
| Product Type | Supertek™ Science Cube Kits |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
