Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Magnetic Fun
GREENER_CHOICESCI_ED

Catalog No. S89517 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89517 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89517 Supplier Supertek™ Supplier No. PH96015S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can you make a magnet float? Magnets have many fascination properties! Learn about them as you make your own magnet, discover what poles are, and make that magnet float. We will get things started with our experiment guide, and then have fun as you discover all there is to learn about magnetism.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 3 magnetic wands
    • 36 magnetic marbles
    • 3 magnetic rings
    • 1 floating magnet stand
    • 1 compass
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Construct a Magnet, Discover Poles, Make a Magnet Float
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.