Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Simple Circuit
GREENER_CHOICESCI_ED

Catalog No. S89513 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89513 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89513 Supplier Supertek™ Supplier No. PH96017S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electricity, we all know what it is; but how does it work? Create a simple circuit as you learn about electricity, currents, voltages, series circuits, and much more.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 1 D cell battery holder
    • 1 knife switch
    • 2 light bulb holders
    • 2 light bulbs
    • 5 alligator connector cords
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Creating a Simple Circuit
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.