Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Telegraph
GREENER_CHOICESCI_ED

Catalog No. S89512 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89512 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89512 Supplier Supertek™ Supplier No. PH96018S3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Discover how people communicated long distances, before the telephone! With our Science Cube Telegraph Kit you can experience a little bit of history, learn Morse Code, and even send a secret message to a friend.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 1 D cell battery holder
    • 1 contact key
    • 2 light bulbs
    • 5 alligator connector cords
    • 1 buzzer
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Group Experiments and Homeschooling
Includes Reusable Plastic Snap Box, D Cell Battery Holder, Contact Key, Light Bulbs, Alligator Connector Cords, Buzzer, Exploration Guide
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.