Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Test Tubes, Rack + More
GREENER_CHOICESCI_ED

Catalog No. S89522 Shop All Supertek Scientific Products
Change view
Click to view available options
Material:
Borosilicate Glass
Polystyrene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S89522 Borosilicate Glass
S89523 Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S89522 Supplier Supertek™ Supplier No. PH96008S3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Get a jumpstart on your next science experiment with Test Tubes Rack More Kits. A great basic kit featuring some of the most common tools found in all labs. The test tubes are made from borosilicate glass or polystyrene.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 6 Test Tubes
    • 2 Pipettes
    • 1 Test Tube Tong
    • 2 Test Tube Brushes
    • 1 Test Tube Holder
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
Material Borosilicate Glass
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.