Learn More
Science Fair Projects

Description
Put together first-rate science fair projects that will reinforce the scientific method while impressing the judges. Books will help students choose and develop their own ideas or suggest some outlines that relate to current topics.
The Complete Handbook of Science Fair Projects.
Newly-revised and updated version of this popular handbook includes 50 award-winning projects and an expanded list of 500 fascinating topics. A great resource for the lastest ISEF rules and guidelines plus information on resources and state or regional science fair listings.
More Award-Winning Science Fair Projects.
A great follow-up to The Complete Handbook of Science Fair Projects, this book features 50 additional award-winning projects, described in detail with accompanying illustrations. List of 500 topic ideas will inspire young scientists. Provides a thorough guide to putting together a science fair project, as well as the latest science fair rules, resources, and useful websites.
Specifications
Specifications
| Format | Book |
| Product Type | The Complete Handbook of Science Fair Projects |
| Includes | 50 Award-winning projects and an expanded list of 500 fascinating topics |
| For Use With (Application) | The classic reference on how to do a science fair project features step-by-step instructions describing 50 award-winning projects from actual science fairs |
| Subject | Science |
| Author | Julianne Blair Bochinski |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.