Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciencell Research Laboratories ScienCell Recombinant Human Serum Albumin, 50 g, ≥ 99% Purity

Catalog No. NC3983797
Encompass

Recombinant Human Serum Albumin (rHSA) is a highly purified, animal-, virus-, and bacteria-free product derived from a rice-based expression system. It serves as a structurally equivalent alternative to plasma-derived HSA. For research use only.

  • Source: Oryza sativa (rice grain)
  • Purity: ≥ 99% by SDS-PAGE
  • Physical appearance: Light yellow to white powder
  • Endotoxin: <0.005EU/μg
  • Storage: Store at 2–8°C; shipped on gel ice
  • Eliminates risks of bacterial and viral contamination; offers superior consistency and economical price.
  • Suitable for cell culture media supplementation, cell cryopreservation, electrophoresis, Western blots, and ELISA.

Catalog No. NC3983797 Supplier Sciencell Research Laboratories Supplier No. OSRHSA50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Human Serum Albumin (rHSA) is a highly purified, animal-, virus-, and bacteria-free product derived from a rice-based expression system. It serves as a structurally equivalent alternative to plasma-derived HSA. For research use only.

  • Source: Oryza sativa (rice grain)
  • Purity: ≥ 99% by SDS-PAGE
  • Physical appearance: Light yellow to white powder
  • Endotoxin: <0.005EU/μg
  • Storage: Store at 2–8°C; shipped on gel ice
  • Eliminates risks of bacterial and viral contamination; offers superior consistency and economical price.
  • Suitable for cell culture media supplementation, cell cryopreservation, electrophoresis, Western blots, and ELISA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.