Learn More
Research Products International Corp Scienceware Orbital Shaker, Vibrating
SDP
Catalog No. 501977174
Encompass
Save the expense and eliminate the space requirements of another piece of lab equipment. Placed and centered on a magnetic stirrer, this platform operates as an orbital vibrator for plates, flasks or other vessels.
Vibrate and mix; rotating orbit diameter of 1.52 mm (1/16 inch)
Vibration adjustment is controlled by setting the magnetic stirrer to the desired speed
Platform protects the operating mechanism from spills; non-skid surface to prevent containers from sliding
Base incorporates built-in hand holds for lifting and carrying
Supports up to 6.8 kg (15 pounds)
Catalog No. 50-197-7174 Supplier Research Products International Corp Supplier No. 437041
Save the expense and eliminate the space requirements of another piece of lab equipment. Placed and centered on a magnetic stirrer, this platform operates as an orbital vibrator for plates, flasks or other vessels.
Vibrate and mix; rotating orbit diameter of 1.52 mm (1/16 inch)
Vibration adjustment is controlled by setting the magnetic stirrer to the desired speed
Platform protects the operating mechanism from spills; non-skid surface to prevent containers from sliding
Base incorporates built-in hand holds for lifting and carrying
Supports up to 6.8 kg (15 pounds)