Learn More
New York Microscope Company Inc SCIENSCOPE NZ-PK3FX-R3, NZ STE
SDP
Catalog No. NC2217381
Encompass
Scienscope NZ-PK3FX-R3, NZ Stereo Zoom Binocular Microscope on Heavy Duty Articulating Arm with LED Ring Light with Polarizer, 8x to 50x Magnification
This item is not returnable. View return policy
Catalog No. NC2217381 Supplier New York Microscope Company Inc Supplier No. SCSNZPK3FXR3
Scienscope NZ-PK3FX-R3, NZ Stereo Zoom Binocular Microscope on Heavy Duty Articulating Arm with LED Ring Light with Polarizer, 8x to 50x Magnification