Learn More
Electron Microscopy Sciences Scientific Calculator 5¼" x 2¾" x 7/16"
SDP
Catalog No. 5028502
Encompass_Preferred
Highly efficient solar panel requires only 50 lux, works with any available light. Will never need batteries. 10 digit LCD + 2 digit exponent. 15 levels of parenthesis. 67 functions including all the preprogrammed functions of Large Display Scientific Calculator 830003 plus; binary, octal decimal, sexagesimal, and hexadecimal conversion, and standard deviation. Comes with a soft carrying pouch, instructions and a button cell battery.
Catalog No. 50-285-02 Supplier Electron Microscopy Sciences Supplier No. 70090
Highly efficient solar panel requires only 50 lux, works with any available light. Will never need batteries. 10 digit LCD + 2 digit exponent. 15 levels of parenthesis. 67 functions including all the preprogrammed functions of Large Display Scientific Calculator 830003 plus; binary, octal decimal, sexagesimal, and hexadecimal conversion, and standard deviation. Comes with a soft carrying pouch, instructions and a button cell battery.