Learn More
Scientific Device Cryptosporidium Control Slides
SDP
Catalog No. 23769266 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
10 slides
Catalog No. Quantity
23-769266 10 slides
1 options
This item is not returnable. View return policy
Catalog No. 23-769266 Supplier Scientific Device Supplier No. 370
This item is not returnable. View return policy
Many typical organisms on each slide
  • Cryptosporidium sps. is now recognized as an important intestinal pathogen in patients with HIV infection
  • Routine parasitological techniques fail to demonstrate this protozoa
  • Modified Acid Fast staining techniques prove effective in staining this organism
  • Each package contains 10 slides, each having a positive and negative well
Specifications
For Use With (Application) Used for staining both cryptosporidium and nocardia
Includes Positive and negative well on each slide
Quantity 10 slides