Learn More
Scientific Device Fecal Concentrator Kits
SDP
Catalog No. 23769508 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
50/Cs.
Catalog No. Quantity
23-769-508 50/Cs.
1 options
Catalog No. 23-769-508 Supplier Scientific Device Supplier No. 641
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
50 standardized fine mesh filter funnels, 50 polypropylene centrifuge tubes of 15mL, 50 tube caps and one 15mL bottle of mucolytic digesting fluid
For concentration of feces for ova and parasites
  • Safe and disposable
  • Contains all components needed for feces concentration
Specifications
Product Type Fecal Concentrator Kit
Includes 50 Standardized fine mesh filter funnels, 50 x 15mL Polypropylene centrifuge tubes, 50 tube caps and 15mL bottle of Mucolytic digesting fluid
Quantity 50/Cs.