Learn More
Scientific Device Fungi-Tape™
SDP
Catalog No. 23769321 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Product Type:
Fungi Tape
Catalog No. Product Type
23-769-321 Fungi Tape
1 options
This item is not returnable. View return policy
Catalog No. 23-769-321 Supplier Scientific Device Supplier No. 745
This item is not returnable. View return policy
Provides an efficient way of performing slide mounts of fungi from primary cultures
  • Pliable enough to make tape loops for sampling
  • Will not disintegrate or bubble
  • Contains approximately 360 coverslips
  • Adhesive
  • Dimensions: 2.54 × 22.9cm (1 in. × 25 ft.)
  • Clear
Compatible with Scientific Device MycoPerm.
Specifications
Product Type Fungi Tape