Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device KOH-Methylene blue Fungal Control Suspension
SDP

Catalog No. 23769515 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-515 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-769-515 Supplier Scientific Device Supplier No. 6025

missing translation for 'validMainframeMsg'

Quality control for potassium hydroxide (KOH) Prep.

  • Contains 1mL suspension of inactivated mycelia and spores from Aspergillus sps. with cellular debris
  • Perfect control for skin scraping

Specifications

Quantity 1 Pack
Includes 1mL suspension of inactivated mycelia and spores from Aspergillus species with cellular debris
Format Vial
For Use With (Application) Skin scraping
Product Type KOH-Methylene Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.