Learn More
Scientific Device Laboratory 2 x 3 Inch Round Well Agglutination Slides
SDP
Catalog No. 23769451 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Round well
Catalog No.
23-769-451 Round well
1 options
This item is not returnable. View return policy
Catalog No. 23-769-451 Supplier Scientific Device Supplier No. 0742
This item is not returnable. View return policy
Specifications
Well Count 1
For Use With (Application) Latex and other agglutination techniques
Length (English) 2 in.
Width (English) 2 in.
Dimensions (L x W) 3 x 2 in.