Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Laboratory Aqua-Hold Pap Pen 2
SDP

Catalog No. 23769533 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-533 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-769-533 Supplier Scientific Device Supplier No. 980402

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Creates liquid repellent area on slides

  • Draws liquid repellent area around section of tissue so that expensive stains and reagents are not wasted
  • Insoluble in alcohol and acetone but can be removed after staining with xylene or SDL's Paraclear™
  • Contains 13mL of barrier material
  • Red tint

Specifications

Product Type Aqua-hold 2 Pap Pen
Color Red
For Use With (Application) Draws liquid repellent area around section of tissue so that expensive stains and reagents are not wasted
Quantity 1 Ea.
Capacity (Metric) 13 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.