Learn More
Scientific Device Laboratory Gram Stain Control Slides
SDP
Catalog No. 23769303 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
50 Slides
Catalog No. Quantity
23-769-303 50 Slides
1 options
Catalog No. 23-769-303 Supplier Scientific Device Laboratory, Inc Supplier No. 361
Specially designed to place clinical smear and control organism on same slide
  • Available pack size is 50 slides/Pk.
  • With space for writing
Specifications
Product Type Control Slide
For Use With (Application) Clinical purpose
Quantity 50 Slides