Learn More
Scientific Device McFarland Standards
SDP
Catalog No. 23769315 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
5 Pack
Catalog No. Quantity
23-769-315 5 Pack
1 options
Catalog No. 23-769-315 Supplier Scientific Device Supplier No. 2350
Available in McFarland or Colorimeter Standard Sets
  • Each particle is the size of Entamoeba Coli
  • Each tube contains 12mL (.40 oz.) of suspension
  • Latex particle suspension
  • Extended shelf life
Specifications
Content And Storage Room Temperature
Includes 0.5, 1, 2, 3 and 4 Standard
Quantity 5 Pack