Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Modified Trichrome Stain Kit for Microsporidia
SDP

Catalog No. 23769519 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-519 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-769-519 Supplier Scientific Device Supplier No. 7843

missing translation for 'validMainframeMsg'

Used for staining of microsporidia

  • Includes one 250mL bottle of stain and one 250mL bottle of decolorizer/counterstain

Specifications

Quantity 1 Kit
Includes 250mL stain and 250mL decolorizer/counterstain
Format Bottle
For Use With (Application) Staining of microsporidia
Product Type Modified Trichrome Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.