Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Paraclear Clearing Agents
SDP

Catalog No. 23769529 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
19000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-529 19000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-769-529 Supplier Scientific Device Supplier No. 6155

missing translation for 'validMainframeMsg'

Xylene-free clearing agent

  • Xylene-free, without overpowering odor
  • No health hazards
  • Evaporates very fast
  • Trichrome procedure
  • Soluble in alcohol, deparaffinized easily; but has no effect on laboratory plastics
  • Less flammable as campared to xylene

Specifications

Boiling Point 36.85°C
Product Type Tissue Clearing Reagent
Quantity 19000mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.