Learn More
Scientific Device Pneumocystis Control Slides
SDP
Catalog No. 23769503 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
23-769-503 10/Pk.
1 options
Catalog No. 23-769-503 Supplier Scientific Device Supplier No. 356
Convenient control slide
  • Contains homogenated lung tissue with many organisms similar to microbiology laboratory samples
  • Quantity: 10 slides/Pk.
These slides should NOT be used with current monoclonal antibody fluorescent assays.
Specifications
Quantity 10/Pk.