Learn More
Scientific Device Printed TB Slides
SDP
Catalog No. 23769520 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
TB Printed Slides, Well size: 20mm
Catalog No.
23-769-520 TB Printed Slides, Well size: 20mm
1 options
This item is not returnable. View return policy
Catalog No. 23-769-520 Supplier Scientific Device Supplier No. 0410120
This item is not returnable. View return policy
Ideal for making smears when mycobacterial disease is suspected
  • Slides with 20mm well provides enough space to accommodates large amount of material
  • Black epoxy slides
  • Used for traditional acid fast or fluorescent staining
Specifications
Type TB Printed Slide
For Use With (Application) For making smears when mycobacterial disease is suspected