Learn More
Scientific Device Shrink Seals
SDP
Catalog No. 23769501 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Type:
Seal, Shrink
Catalog No. Type
23-769-501 Seal, Shrink
1 options
Catalog No. 23-769-501 Supplier Scientific Device Supplier No. 261
For preparing in-house plates which requires prolonged incubation
  • Provide safe sealing to petri-dishes and test-tubes
  • Ideal for preparing sensitive plates for TB and fungus culture which are not used immediately
  • Colorless cellulose bands
  • Easy to apply
  • Stacking blocks enables to dry multiple plates in lesser space
  • Standard size: 92 x 25mm
Specifications
Type Seal, Shrink
For Use With (Application) For safe sealing of petri-dishes or test-tubes