Learn More
Scientific Device Sterile Instruments pack with cutting board
SDP
Catalog No. 23769512 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
10 Sets/Pk.
Catalog No. Quantity
23-769-512 10 Sets/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-769-512 Supplier Scientific Device Supplier No. 906
This item is not returnable. View return policy
Ideal for lab work where sterile instruments are needed very frequently at short notice
  • Pre-packaged and ready-to-use
  • Disposable nature avoid washing and possible contamination
  • Includes sterile disposable dissection board, fine-tipped forceps, One rigid plastic scalpel holder with no. 11 blade and one stainless steel pointed scissors
  • Autoclavable Scissors and forceps
  • 10 sets/Pk.
Specifications
Product Type Sterile Instrument Pack
Disposable Single-use
For Use With (Application) Laboratory use
Quantity 10 Sets/Pk.