Learn More
Cole-Parmer Idex P-200X Flangeless Ferrule, Blue ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Catalog No. NC1672005 Shop All Cole-Parmer Products
Encompass
Use with male nuts for a leak-tight connection.
Catalog No. NC1672005 Supplier Cole-Parmer Supplier No. 0193930
Use with male nuts for a leak-tight connection.