Learn More
AB Sciex LLC Sciex Dual Tap Transformer, 194-220VAC Supply, 2 Internal Taps, 1 Ea
Catalog No. NC3947206
Encompass_Preferred
This dual tap boost transformer is designed to regulate power supply in environments where the input voltage ranges between 194-220VAC. It facilitates consistent operation by allowing for voltage adjustment through internal configurations.
  • Designed for supply voltage between 194-220VAC
  • Features two internal taps for voltage selection
Catalog No. NC3947206 Supplier AB Sciex LLC Supplier No. 5029592Q220567
This dual tap boost transformer is designed to regulate power supply in environments where the input voltage ranges between 194-220VAC. It facilitates consistent operation by allowing for voltage adjustment through internal configurations.
  • Designed for supply voltage between 194-220VAC
  • Features two internal taps for voltage selection