Learn More
Scigen Neutra Wipes™ Dispenser
Catalog No. 23730566 Shop All Scigen Inc Products
Change view
Click to view available options
Packaging Quantity:
60/Pk.
Catalog No. Packaging Quantity
23-730-566 60/Pk.
1 options
Catalog No. 23-730-566 Supplier Scigen Supplier No. 4278
Add to Cart
Add to Cart
Includes
Dispenser and 1 roll of wipes.
Rapidly absorb and neutralize small formalin spills. With convenient dispenser.
  • Convenient pop-up dispenser
  • Absorbs and neutralizes noxious vapors
  • 60 wipes (12 x 12.5 in.) per roll
  • Manufactured using ISO 13485 quality system
  • Ideal as liners on storage carts, shelves, and in rooms where formalin and aldehyde are kept or transported—neutralizing any spillage and helping to eliminate noxious vapors
  • Designed specifically for use with other Neutralex products for maximum effectiveness in treating formalin and aldehyde waste
Note: For convenient wipe refills, see Neutra-Wipes - Refill Rolls (4279K1 and 4279)
Specifications
Type Neutra Wipes™
For Use With (Application) Absorbs and neutralizes small formalin spills
Includes Dispenser and 1 roll of 60 wipes
Packaging Quantity 60/Pk.