Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab SciGen Neutralex Neutralizer
SDP

Catalog No. 21402140
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-140 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-140 Supplier StatLab Supplier No. CWS4047
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Provides complete aldehyde neutralization in 15 minutes and is a CAL/EPA certified technology

  • Uses an anti-drain-clogging polymer by-product that brings final neutralization levels down to 10 ppm
  • Requires no pH adjustment and is a pre-measured powder in easy-to-use pouches
  • Each pouch neutralizes 1 gallon of 10% Formalin
  • 16 pouches per case

Specifications

Physical Form Solid
Color Colorless
Quantity 0.075 kg
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.