Learn More
Scilogex LLC SCI-RL-Pro LCD Digital Rotisserie Tube Rotator
Supplier:  Scilogex LLC 824222019999
Encompass_Preferred
The SCI-RL-PRO provides a gentle but effective mixing of biological samples in 1.5ml to 50ml micro tubes, used in a variety of applications including immune precipitations, prevention of blood coagulation, latex diagnostics etc.
Catalog No. NC1541949