Learn More
Scilogex LLC Linear Pipettor Stand, holds 6 MicroPette Pipettors
Catalog No. NC0694852
Encompass_Preferred
Scilogex Linear Pipettor Stand For 6 Pipettors The Linear And Round Pipette Stands Have Been Designed To Fit Perfectly The Scilogex Pipette Range: Toppette, Micropette And Micropette Plus Autoclavable Pipettes. The Stands Are Convenient To Hold Up 6 Pipettes Both Of Single And Multi-Channel Units.
Catalog No. NC0694852 Supplier Scilogex LLC Supplier No. 710000859999
Scilogex Linear Pipettor Stand For 6 Pipettors The Linear And Round Pipette Stands Have Been Designed To Fit Perfectly The Scilogex Pipette Range: Toppette, Micropette And Micropette Plus Autoclavable Pipettes. The Stands Are Convenient To Hold Up 6 Pipettes Both Of Single And Multi-Channel Units.