Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ScintiGest™ Tissue Solubilizer, 0.6N (Scintanalyzed™), Fisher Chemical

Catalog No. SX10100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SX10-100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SX10-100

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 65°C
Chemical Name or Material ScintiGest™ Tissue Solubilizer
Name Note 0.6N
Physical Form Liquid
CAS 57-09-0, 67-56-1, 1310-58-3
Color Yellow
Quantity 100 mL
MDL Number MFCD00212665
LSC Analysis Pass Test
Packaging Glass Bottle
Grade Scintanalyzed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.