Learn More
CHEMSCIENCE Scintilation Vial, 20mL, comes with Polypropylene cap, Pulp-Backed Foil Liner
Catalog No. NC2117306
Encompass_Preferred
Scintilation Vial, 20mL, comes with Polypropylene cap, Pulp-Backed Foil Liner
This item is not returnable. View return policy
Catalog No. NC2117306 Supplier CHEMSCIENCE Supplier No. CS100020
Scintilation Vial, 20mL, comes with Polypropylene cap, Pulp-Backed Foil Liner